Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Glu(OBzl)-OH | 5680-86-4 | 99.1% | 371.38 | 100 G
SDP

Catalog No. 5000451647
Encompass_Preferred

Z-Glu(OBzl)-OH is a glutamic acid derivative used as a laboratory chemical and for the manufacture of substances. It is stable under recommended storage conditions.

  • Glutamic acid derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Beneficial as an ergogenic dietary substance
  • Appearance: White to off-white solid
  • Melting/freezing point: 74.0-78.0 °C
  • Boiling point/range: 594.3°C at 760 mmHg
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only
  • Not fully validated for medical applications

Catalog No. 50-004-51647 Supplier Medchemexpress LLC Supplier No. HYW011074100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Glu(OBzl)-OH is a glutamic acid derivative used as a laboratory chemical and for the manufacture of substances. It is stable under recommended storage conditions.

  • Glutamic acid derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Beneficial as an ergogenic dietary substance
  • Appearance: White to off-white solid
  • Melting/freezing point: 74.0-78.0 °C
  • Boiling point/range: 594.3°C at 760 mmHg
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only
  • Not fully validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.