Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Glu-OtBu | 5891-45-2 | 99.65% | 337.37 | 10 G
SDP

Catalog No. 5000395581
Encompass_Preferred

Z-Glu-OtBu is a glutamic acid derivative primarily utilized as a laboratory chemical in the manufacture of various substances. It presents as a white to off-white solid, offering high purity and stability for diverse research applications. Its precise molecular structure and composition are well-defined, ensuring reliability in experimental contexts.

  • High purity of 99.65% (HPLC)
  • White to off-white solid appearance
  • Molecular formula C17H23NO6
  • Molecular weight 337.37
  • Stable under recommended storage conditions

Catalog No. 50-003-95581 Supplier Medchemexpress LLC Supplier No. HYW00854910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Glu-OtBu is a glutamic acid derivative primarily utilized as a laboratory chemical in the manufacture of various substances. It presents as a white to off-white solid, offering high purity and stability for diverse research applications. Its precise molecular structure and composition are well-defined, ensuring reliability in experimental contexts.

  • High purity of 99.65% (HPLC)
  • White to off-white solid appearance
  • Molecular formula C17H23NO6
  • Molecular weight 337.37
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.