Learn More
Medchemexpress LLC Z-Glu-OtBu | 5891-45-2 | 99.7% | 337.37 | 100 G
SDP
Catalog No. 5000395580
Encompass_Preferred
Z-Glu-OtBu (HY-W008549) is a chemical substance supplied by MedChemExpress for laboratory research and the manufacture of other substances. This product appears as a white to off-white solid and is stable under recommended storage conditions.
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Ships at room temperature if less than 2 weeks
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Complies with given specifications
  • For research use only
Catalog No. 50-003-95580 Supplier Medchemexpress LLC Supplier No. HYW008549100G
Z-Glu-OtBu (HY-W008549) is a chemical substance supplied by MedChemExpress for laboratory research and the manufacture of other substances. This product appears as a white to off-white solid and is stable under recommended storage conditions.
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Ships at room temperature if less than 2 weeks
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Complies with given specifications
  • For research use only