Learn More
Medchemexpress LLC Z-Glu-Tyr-OH | 988-75-0 | 99.4% | 25 MG
SDP
Catalog No. 5000341706
Encompass_Preferred
Z-Glu-Tyr-OH is a peptide used for the synthesis of peptides on a solid support. It appears as a white to off-white solid and is intended for research use only.
  • Can be used for peptide synthesis on a solid support.
  • Appears as a solid, white to off-white in color.
  • For research use only.
  • Store in sealed packaging, away from moisture.
  • Powder stability: 2 years at -80°C, 1 year at -20°C.
  • In solvent stability: 6 months at -80°C, 1 month at -20°C.
Catalog No. 50-003-41706 Supplier Medchemexpress LLC Supplier No. 25MGHY131095
Z-Glu-Tyr-OH is a peptide used for the synthesis of peptides on a solid support. It appears as a white to off-white solid and is intended for research use only.
  • Can be used for peptide synthesis on a solid support.
  • Appears as a solid, white to off-white in color.
  • For research use only.
  • Store in sealed packaging, away from moisture.
  • Powder stability: 2 years at -80°C, 1 year at -20°C.
  • In solvent stability: 6 months at -80°C, 1 month at -20°C.