Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Glu-Tyr-OH | 988-75-0 | 99.4% | 50 MG
SDP

Catalog No. 5000341018
Encompass_Preferred

Z-Glu-Tyr-OH is a compound intended for research use only. It can be utilized for the synthesis of peptides on a solid support.

  • Can be used for the synthesis of peptides on a solid support
  • Intended for research use only
  • Purity of 99.42%
  • Molecular weight is 444.43 g/mol
  • Chemical formula is C22H24N2O8
  • Appearance is a white to off-white solid
  • Ships at room temperature in continental US
  • Requires sealed storage, away from moisture
  • Powder can be stored at -80°C for 2 years or -20°C for 1 year
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-41018 Supplier Medchemexpress LLC Supplier No. HY13109550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Glu-Tyr-OH is a compound intended for research use only. It can be utilized for the synthesis of peptides on a solid support.

  • Can be used for the synthesis of peptides on a solid support
  • Intended for research use only
  • Purity of 99.42%
  • Molecular weight is 444.43 g/mol
  • Chemical formula is C22H24N2O8
  • Appearance is a white to off-white solid
  • Ships at room temperature in continental US
  • Requires sealed storage, away from moisture
  • Powder can be stored at -80°C for 2 years or -20°C for 1 year
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.