Learn More
Medchemexpress LLC Z-Gly-Gly-Arg-AMC | 66216-78-2 | 99.9% | 579.60 | 5 MG
SDP
Catalog No. 5000426604
Encompass_Preferred
Z-Gly-Gly-Arg-AMC is a thrombin-specific fluorogenic substrate used for testing thrombin generation in PRP (platelet-rich plasma) and PPP (platelet-poor plasma), with excitation/emission wavelengths of 390/480 nm. It is intended for research use only.
  • Thrombin-specific fluorogenic substrate
  • Used for testing thrombin generation in PRP and PPP
  • Excitation/emission wavelengths of 390/480 nm
  • Intended for research use only
Catalog No. 50-004-26604 Supplier Medchemexpress LLC Supplier No. HYP00195MG
Z-Gly-Gly-Arg-AMC is a thrombin-specific fluorogenic substrate used for testing thrombin generation in PRP (platelet-rich plasma) and PPP (platelet-poor plasma), with excitation/emission wavelengths of 390/480 nm. It is intended for research use only.
  • Thrombin-specific fluorogenic substrate
  • Used for testing thrombin generation in PRP and PPP
  • Excitation/emission wavelengths of 390/480 nm
  • Intended for research use only