Learn More
Medchemexpress LLC Z-Gly-Gly-Arg-AMC | 66216-78-2 | 99.9% | 579.60 | 50 MG
SDP
Catalog No. 5000426376
Encompass_Preferred
Z-Gly-Gly-Arg-AMC is a thrombin-specific fluorogenic substrate used for testing thrombin generation in PRP and platelet-poor plasma (PPP) (Ex/Em = 390/480 nm).
  • Thrombin-specific fluorogenic substrate.
  • Suitable for testing thrombin generation in PRP and platelet-poor plasma (PPP).
  • Excitation/Emission wavelengths: 390/480 nm.
Catalog No. 50-004-26376 Supplier Medchemexpress LLC Supplier No. HYP001950MG
Z-Gly-Gly-Arg-AMC is a thrombin-specific fluorogenic substrate used for testing thrombin generation in PRP and platelet-poor plasma (PPP) (Ex/Em = 390/480 nm).
  • Thrombin-specific fluorogenic substrate.
  • Suitable for testing thrombin generation in PRP and platelet-poor plasma (PPP).
  • Excitation/Emission wavelengths: 390/480 nm.