Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Gly-Gly-Arg-AMC acetate | 2070009-61-7 | 99.8% | 639.66 | 10 MG
SDP

Catalog No. 501933993
Encompass_Preferred

Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate. It is used for testing thrombin generation in platelet-rich plasma (PRP) and platelet-poor plasma (PPP), with excitation/emission wavelengths of 390/480 nm. This product is intended for research use only.

  • Thrombin-specific fluorogenic substrate
  • Used for testing thrombin generation in platelet-rich plasma and platelet-poor plasma
  • Excitation wavelength 390 nm, emission wavelength 480 nm
  • Solid appearance, white to off-white color
  • Soluble in DMSO at 100 mg/mL and in water at 20 mg/mL
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture and light

Catalog No. 50-193-3993 Supplier Medchemexpress LLC Supplier No. HYP0019A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate. It is used for testing thrombin generation in platelet-rich plasma (PRP) and platelet-poor plasma (PPP), with excitation/emission wavelengths of 390/480 nm. This product is intended for research use only.

  • Thrombin-specific fluorogenic substrate
  • Used for testing thrombin generation in platelet-rich plasma and platelet-poor plasma
  • Excitation wavelength 390 nm, emission wavelength 480 nm
  • Solid appearance, white to off-white color
  • Soluble in DMSO at 100 mg/mL and in water at 20 mg/mL
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.