Learn More
Medchemexpress LLC Z-Gly-Gly-Arg-AMC acetate | 2070009-61-7 | 99.8% | 639.66 | 5 MG
SDP
Catalog No. 5000426696
Encompass_Preferred
Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate used for testing thrombin generation in platelet-rich plasma (PRP) and platelet-poor plasma (PPP). This compound exhibits excitation/emission wavelengths of 390/480 nm.
  • Thrombin-specific fluorogenic substrate
  • Used for testing thrombin generation in PRP and PPP
  • Excitation wavelength of 390 nm
  • Emission wavelength of 480 nm
  • For research use only
Catalog No. 50-004-26696 Supplier Medchemexpress LLC Supplier No. HYP0019A5MG
Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate used for testing thrombin generation in platelet-rich plasma (PRP) and platelet-poor plasma (PPP). This compound exhibits excitation/emission wavelengths of 390/480 nm.
  • Thrombin-specific fluorogenic substrate
  • Used for testing thrombin generation in PRP and PPP
  • Excitation wavelength of 390 nm
  • Emission wavelength of 480 nm
  • For research use only