Learn More
Medchemexpress LLC Z-Gly-Gly-Arg-AMC acetate | 2070009-61-7 | 99.84% | 639.66 | 50 MG
SDP
Catalog No. 5000426605
Encompass_Preferred
Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate primarily used for testing thrombin generation in both platelet-rich plasma (PRP) and platelet-poor plasma (PPP). This compound, characterized by excitation/emission wavelengths of 390/480 nm, is provided as a solid, typically white to off-white, and is strictly for research applications.
  • Thrombin-specific fluorogenic substrate
  • Suitable for thrombin generation assays in PRP and PPP
  • Exhibits excitation at 390 nm and emission at 480 nm
  • Supplied as a white to off-white solid
  • Intended for research use only
Catalog No. 50-004-26605 Supplier Medchemexpress LLC Supplier No. HYP0019A50MG
Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate primarily used for testing thrombin generation in both platelet-rich plasma (PRP) and platelet-poor plasma (PPP). This compound, characterized by excitation/emission wavelengths of 390/480 nm, is provided as a solid, typically white to off-white, and is strictly for research applications.
  • Thrombin-specific fluorogenic substrate
  • Suitable for thrombin generation assays in PRP and PPP
  • Exhibits excitation at 390 nm and emission at 480 nm
  • Supplied as a white to off-white solid
  • Intended for research use only