Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Gly-Pro-AMC | 68542-93-8 | 99.8% | 463.48 g/mol | C25H25N3O6 | 10 MG
SDP

Catalog No. 5000297159
Encompass_Preferred

Z-Gly-Pro-AMC is a fluorogenic peptide substrate used to measure prolyl endopeptidase activity. Enzymatic hydrolysis releases the fluorescent 7-amino-4-methylcoumarin (AMC), enabling sensitive fluorometric assays.

  • Hydrolyzed by prolyl endopeptidase to release 7-amino-4-methylcoumarin (AMC).
  • Excitation 380 nm, emission 465 nm for fluorometric detection.
  • High purity: 99.8%.
  • Molecular weight 463.48 g/mol; molecular formula C25H25N3O6.
  • Available as a 10 mg vial for laboratory assays.
  • Suitable for enzymatic activity assays and fluorescence-based screening.

Catalog No. 50-002-97159 Supplier Medchemexpress LLC Supplier No. HYD167010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Gly-Pro-AMC is a fluorogenic peptide substrate used to measure prolyl endopeptidase activity. Enzymatic hydrolysis releases the fluorescent 7-amino-4-methylcoumarin (AMC), enabling sensitive fluorometric assays.

  • Hydrolyzed by prolyl endopeptidase to release 7-amino-4-methylcoumarin (AMC).
  • Excitation 380 nm, emission 465 nm for fluorometric detection.
  • High purity: 99.8%.
  • Molecular weight 463.48 g/mol; molecular formula C25H25N3O6.
  • Available as a 10 mg vial for laboratory assays.
  • Suitable for enzymatic activity assays and fluorescence-based screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.