Learn More
Medchemexpress LLC Z-Gly-Pro-Arg-AMC hydrochloride | 201928-42-9 | 99.15% | 656.13 | 5 MG
SDP
Catalog No. 5000399733
Encompass_Preferred
Z-Gly-Pro-Arg-AMC hydrochloride is a fluorescent trypsin and cathepsin K substrate. It is used to determine trypsin and cathepsin K activity.
  • Fluorescent trypsin and cathepsin K substrate
  • Can be used to determine trypsin and cathepsin K activity
  • For research use only
Catalog No. 50-003-99733 Supplier Medchemexpress LLC Supplier No. HYP42175MG
Z-Gly-Pro-Arg-AMC hydrochloride is a fluorescent trypsin and cathepsin K substrate. It is used to determine trypsin and cathepsin K activity.
  • Fluorescent trypsin and cathepsin K substrate
  • Can be used to determine trypsin and cathepsin K activity
  • For research use only