Learn More
Medchemexpress LLC Z-Gly-Pro-Arg-AMC hydrochloride | 201928-42-9 | 99.2% | 656.13 | 10 MG
SDP
Catalog No. 5000399583
Encompass_Preferred
Z-Gly-Pro-Arg-AMC hydrochloride is a fluorescent substrate designed for the determination of trypsin and cathepsin K activity.
  • Fluorescent substrate
  • Determines trypsin and cathepsin K activity
Catalog No. 50-003-99583 Supplier Medchemexpress LLC Supplier No. HYP421710MG
Z-Gly-Pro-Arg-AMC hydrochloride is a fluorescent substrate designed for the determination of trypsin and cathepsin K activity.
  • Fluorescent substrate
  • Determines trypsin and cathepsin K activity