Learn More
Bachem Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
SDP
Catalog No. NC9659668
Encompass_Preferred
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH, 13075-38-2, C27H36N6O9, Mr 588.62, 500mg. Substrate for microbial collagenase (MMP-1). Enzymatic cleavage of Z-GPGGPA yields Gly-Pro-Ala, which is determined colorimetrically by reaction with ninhydrin. Z-GPGGPA can induce metamorphosis in larvae and asexual buds of the Cassiopea species of jellyfish in vitro at micromolar concentration.
Catalog No. NC9659668 Supplier Bachem Supplier No. 4002149.0500
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH, 13075-38-2, C27H36N6O9, Mr 588.62, 500mg. Substrate for microbial collagenase (MMP-1). Enzymatic cleavage of Z-GPGGPA yields Gly-Pro-Ala, which is determined colorimetrically by reaction with ninhydrin. Z-GPGGPA can induce metamorphosis in larvae and asexual buds of the Cassiopea species of jellyfish in vitro at micromolar concentration.