Learn More
Bachem Z-Gly-Pro-pNA
SDP
Supplier:  Bachem 4003649.0250
Encompass_Preferred
Z-Gly-Pro-pNA, 65022-15-3, C21H22N4O6, Mr 426.43, 250mg. Z-GP-pNA, chromogenic substrate for post-proline cleaving enzyme (prolyl endopeptidase).
Catalog No. 50-260-391