Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Gly-Pro-pNA | 65022-15-3 | 99.4% | 426.42 | 1 ML
SDP

Catalog No. 5000350318
Encompass_Preferred

Z-Gly-Pro-pNA is a substrate used for measuring prolyl endopeptidase (PEP) inhibitory activity.

  • Substrate for measuring prolyl endopeptidase (PEP) inhibitory activity
  • Molecular weight: 426.42
  • Purity: 99.38%
  • Formula: C21H22N4O6
  • For research use only

Catalog No. 50-003-50318 Supplier Medchemexpress LLC Supplier No. HY13722010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Gly-Pro-pNA is a substrate used for measuring prolyl endopeptidase (PEP) inhibitory activity.

  • Substrate for measuring prolyl endopeptidase (PEP) inhibitory activity
  • Molecular weight: 426.42
  • Purity: 99.38%
  • Formula: C21H22N4O6
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.