Learn More
Enzo Life Sciences Z-GPR-AMC (20mg)
SDP
Catalog No. 502011253
Encompass_Preferred
Fluorogenic substrate specific for cathepsin K, among other cathepsin family members. This substrate is also cleaved by granzyme A, thrombin, tryptase, trypsin, and kallikreins. Ex.: 365 nm, Em.: 440 nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-Gly-Pro-Arg-AMC. Purity: ≥97% (HPLC). Formulation: C31H37N7O7. Long Term Storage: -20°C.
Catalog No. 50-201-1253 Supplier Enzo Life Sciences Supplier No. BMLP1420020
Fluorogenic substrate specific for cathepsin K, among other cathepsin family members. This substrate is also cleaved by granzyme A, thrombin, tryptase, trypsin, and kallikreins. Ex.: 365 nm, Em.: 440 nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-Gly-Pro-Arg-AMC. Purity: ≥97% (HPLC). Formulation: C31H37N7O7. Long Term Storage: -20°C.