Learn More
Cayman Chemical Z-GUGgulsteron 10mg
SDP
Catalog No. 502894088
Encompass_Preferred
A competitive antagonist of farnesoid X receptor (IC50 = 17 µM ) that lowers LDL cholesterol and triglyceride levels in rodents fed a high cholesterol diet; demonstrates antitumor-promoting effects in human multiple myeloma and DU145 human prostate cancer cells
Catalog No. 50-289-4088 Supplier Cayman Chemical Supplier No. 7180010 MG
A competitive antagonist of farnesoid X receptor (IC50 = 17 µM ) that lowers LDL cholesterol and triglyceride levels in rodents fed a high cholesterol diet; demonstrates antitumor-promoting effects in human multiple myeloma and DU145 human prostate cancer cells