Learn More
Medchemexpress LLC (Z)-Guggulsterone | 39025-23-5 | 99.4% | 312.45 | 10 MG
SDP
Catalog No. 5000364886
Encompass_Preferred
(Z)-Guggulsterone is a constituent of the Indian Ayurvedic medicinal plant *Commiphora mukul*. It inhibits the growth of human prostate cancer cells by causing apoptosis and inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis. It also acts as a potent FXR antagonist and reduces ACE2 expression and SARS-CoV-2 infection.
  • Inhibits growth of human prostate cancer cells by causing apoptosis
  • Inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis
  • Acts as a potent FXR antagonist
  • Reduces ACE2 expression
  • Reduces SARS-CoV-2 infection
Catalog No. 50-003-64886 Supplier Medchemexpress LLC Supplier No. 10MGHY110066
(Z)-Guggulsterone is a constituent of the Indian Ayurvedic medicinal plant *Commiphora mukul*. It inhibits the growth of human prostate cancer cells by causing apoptosis and inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis. It also acts as a potent FXR antagonist and reduces ACE2 expression and SARS-CoV-2 infection.
  • Inhibits growth of human prostate cancer cells by causing apoptosis
  • Inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis
  • Acts as a potent FXR antagonist
  • Reduces ACE2 expression
  • Reduces SARS-CoV-2 infection