Learn More
Medchemexpress LLC (Z)-Guggulsterone | 39025-23-5 | 99.4% | 312.45 | 25 MG
SDP
Catalog No. 5000347406
Encompass_Preferred
(Z)-Guggulsterone, a constituent of the Indian Ayurvedic medicinal plant Commiphora mukul, has been shown to inhibit the growth of human prostate cancer cells by inducing apoptosis. It also inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis. Furthermore, (Z)-Guggulsterone acts as a potent FXR antagonist and can reduce ACE2 expression and SARS-CoV-2 infection.
  • Inhibits growth of human prostate cancer cells by inducing apoptosis.
  • Inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis.
  • Acts as a potent FXR antagonist.
  • Reduces ACE2 expression and SARS-CoV-2 infection.
Catalog No. 50-003-47406 Supplier Medchemexpress LLC Supplier No. 25MGHY110066
(Z)-Guggulsterone, a constituent of the Indian Ayurvedic medicinal plant Commiphora mukul, has been shown to inhibit the growth of human prostate cancer cells by inducing apoptosis. It also inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis. Furthermore, (Z)-Guggulsterone acts as a potent FXR antagonist and can reduce ACE2 expression and SARS-CoV-2 infection.
  • Inhibits growth of human prostate cancer cells by inducing apoptosis.
  • Inhibits angiogenesis by suppressing the VEGF-VEGF-R2-Akt signaling axis.
  • Acts as a potent FXR antagonist.
  • Reduces ACE2 expression and SARS-CoV-2 infection.