Learn More
Enzo Life Sciences Z-Guggulsterone (5mg). CAS: 39025-23-5
SDP
Supplier:  Enzo Life Sciences BMLGR2350005
Encompass_Preferred
A guggul tree steroid well known for its lipid lowering properties. Acts as a selective antagonist at farnesoid X receptors (FXRs). Inhibits FXR transactivation (IC50=10µM). Purity: ≥98%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0943