Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-hexadec-9-enamide (palmitoleamide) | 106010-22-4 | MFCD07374311 | 95.0% | 253.42 | C16H31NO | 10 MG
SDP

Catalog No. 5000281663
Encompass_Preferred

(Z)-Hexadec-9-enamide (palmitoleamide) is a fatty acid-derived primary amide used as a research reagent. It is identified by CAS 106010-22-4, has molecular formula C16H31NO and molecular weight 253.42, and is supplied for laboratory research use only.

  • Primary fatty amide derived from palmitoleic acid.
  • Molecular formula C16H31NO, molecular weight 253.42.
  • CAS number 106010-22-4 for unambiguous identification.
  • Supplied as 10 MG solid with listed purity of 95.0%.
  • For laboratory research use only; not for human or clinical use.

Catalog No. 50-002-81663 Supplier Medchemexpress LLC Supplier No. HYW39816610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-Hexadec-9-enamide (palmitoleamide) is a fatty acid-derived primary amide used as a research reagent. It is identified by CAS 106010-22-4, has molecular formula C16H31NO and molecular weight 253.42, and is supplied for laboratory research use only.

  • Primary fatty amide derived from palmitoleic acid.
  • Molecular formula C16H31NO, molecular weight 253.42.
  • CAS number 106010-22-4 for unambiguous identification.
  • Supplied as 10 MG solid with listed purity of 95.0%.
  • For laboratory research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.