Learn More
Medchemexpress LLC Z-IETD-AFC | 219138-02-0 | 99.74% | 821.75 | 10 MG
SDP
Catalog No. 5000422184
Encompass_Preferred
Z-IETD-AFC is a specific fluorescence substrate that can be used to determine caspase-8 catalytic activity.
  • Specific fluorescence substrate
  • Determines caspase-8 catalytic activity
  • Appears as a white to off-white solid
  • Has an emission wavelength of 500
  • Has an excitation wavelength of 380
  • Soluble in DMSO at concentrations greater than or equal to 100 mg/mL
Catalog No. 50-004-22184 Supplier Medchemexpress LLC Supplier No. HYP220810MG
Z-IETD-AFC is a specific fluorescence substrate that can be used to determine caspase-8 catalytic activity.
  • Specific fluorescence substrate
  • Determines caspase-8 catalytic activity
  • Appears as a white to off-white solid
  • Has an emission wavelength of 500
  • Has an excitation wavelength of 380
  • Soluble in DMSO at concentrations greater than or equal to 100 mg/mL