Learn More
Medchemexpress LLC HY-101297 1mg Medchemexpress, Z-IETD-FMK CAS:210344-98-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012971MG
Encompass_Preferred
Medchemexpress, HY-101297 1mg Z-IETD-FMK CAS:210344-98-2 Z-IETD-FMK is a selective caspase-8 inhibitor. Z-IETD-FMK is a granzyme B inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7826