Learn More
Abcam Z-IETD-FMK, Caspase-8 inhibitor, 1MG
Catalog No. 502706658
Encompass_Preferred
MW 654.7 Da, Purity >99%. Z-IETD-FMK, Caspase-8 inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6658 Supplier Abcam Supplier No. AB1413821MG
MW 654.7 Da, Purity >99%. Z-IETD-FMK, Caspase-8 inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement