Learn More
Enzo Life Sciences Z-IETD-FMK (Ready-to-Use) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX260144R100
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-8 and granzyme B. Inhibits also caspase-10. The inhibitor is designed as a methyl ester to facilitate cell permeability.  Alternative name: Caspase 8 inhibitor (fluoromethylketone), Granzyme B inhibitor (fluoromethylketone). MW: 654.0. Purity: ≥99% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8353