Learn More
CPC Scientific (Z-Ile-Glu-Thr-Asp)2-R110 0.5MG
SDP
Catalog No. 502109070
Encompass
ONE-LETTER SEQUENCE: (Z-IETD)2-R110MOLECULAR FORMULA: C74H86N10O25MOLECULAR WEIGHT:1515.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Science 281, 1312 (1998); J Biol Chem 276, 6974 (2001); Biochemistry 38, 13906 (1999)
Catalog No. 50-210-9070 Supplier CPC Scientific Supplier No. CASP080A
ONE-LETTER SEQUENCE: (Z-IETD)2-R110MOLECULAR FORMULA: C74H86N10O25MOLECULAR WEIGHT:1515.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Science 281, 1312 (1998); J Biol Chem 276, 6974 (2001); Biochemistry 38, 13906 (1999)