Learn More
CPC Scientific Z-Ile-Glu-Thr-Asp-AFC 5MG
SDP
Supplier:  CPC Scientific CASP064B
Encompass
Fluorogenic substrate that corresponds to one of the cleavage sites of the inactive 32 kD caspase-3 precursor (amino acids 172-175). Substrate for granzyme B.ONE-LETTER SEQUENCE: Z-IETD-AFCMOLECULAR FORMULA: C37H42N5O13F3MOLECULAR WEIGHT:821.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [219138-02-0], RESEARCH AREA: ApoptosisREFERENCES: Z. Han et al., J. Biol. Chem., 272, 13432 (1997)
Catalog No. 50-210-9034