Learn More
Medchemexpress LLC Z-Ile-Leu-aldehyde | 161710-10-7 | MFCD04974201 | 97.3% | 362.46 g/mol | C20H30N2O4 | 10MG
SDP
Catalog No. 5000237126
Encompass_Preferred
Z-Ile-Leu-aldehyde (Z-IL-CHO) is a peptide aldehyde that competitively inhibits γ-secretase and Notch signaling. It is used in cell-based and animal studies to probe Notch-dependent pathways, induce apoptosis in Notch-active myeloma cells, and reduce osteolytic lesions in preclinical models. The compound is supplied as a white to off-white solid with high purity and is soluble in DMSO for assay preparation.
Catalog No. 50-002-37126 Supplier Medchemexpress LLC Supplier No. HY1246510MG
Z-Ile-Leu-aldehyde (Z-IL-CHO) is a peptide aldehyde that competitively inhibits γ-secretase and Notch signaling. It is used in cell-based and animal studies to probe Notch-dependent pathways, induce apoptosis in Notch-active myeloma cells, and reduce osteolytic lesions in preclinical models. The compound is supplied as a white to off-white solid with high purity and is soluble in DMSO for assay preparation.