Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Ile-Leu-aldehyde | 161710-10-7 | MFCD04974201 | 97.3% | 362.46 g/mol | C20H30N2O4 | 10MM 1ML
SDP

Catalog No. 5000238809
Encompass_Preferred

Z-Ile-Leu-aldehyde (Z-IL-CHO) is a peptide aldehyde inhibitor of γ-secretase and Notch signaling used in biochemical and cell-based research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid material in multiple milligram quantities for assay preparation and dose-response studies.

  • Potent competitive peptide aldehyde inhibitor of γ-secretase and Notch.
  • Available as 10 mM solution in DMSO and as solid in 1 mg-100 mg sizes.
  • Molecular formula C20H30N2O4; molecular weight 362.46 g/mol.
  • Purity 97.3% as reported by manufacturer.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended stability.

Catalog No. 50-002-38809 Supplier Medchemexpress LLC Supplier No. HY1246510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Ile-Leu-aldehyde (Z-IL-CHO) is a peptide aldehyde inhibitor of γ-secretase and Notch signaling used in biochemical and cell-based research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid material in multiple milligram quantities for assay preparation and dose-response studies.

  • Potent competitive peptide aldehyde inhibitor of γ-secretase and Notch.
  • Available as 10 mM solution in DMSO and as solid in 1 mg-100 mg sizes.
  • Molecular formula C20H30N2O4; molecular weight 362.46 g/mol.
  • Purity 97.3% as reported by manufacturer.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.