Learn More
Medchemexpress LLC (Z)-JQ1-TCO | 2087490-43-3 | 99.3% | 609.18 | 100 MG
SDP
Catalog No. 5000359522
Encompass_Preferred
JQ1-TCO (JQ1-trans-cyclooctene) is a derivative of JQ1, an inhibitor of BET. This compound is suitable for click chemistry and can be used as molecular probes in vitro and in vivo. Studies indicate that (Z)-JQ1-TCO does not induce significant degradation of BRD4 in HeLa cells at 10 μM for 18 h.
  • Derivative of JQ1, an inhibitor of BET
  • Suitable for click chemistry
  • Can be used as molecular probes in vitro and in vivo
  • Does not induce significant degradation of BRD4 in HeLa cells at 10 μM for 18 h
  • Targets BRD4 and the Epigenetic Reader Domain
Catalog No. 50-003-59522 Supplier Medchemexpress LLC Supplier No. HY148864A100MG
JQ1-TCO (JQ1-trans-cyclooctene) is a derivative of JQ1, an inhibitor of BET. This compound is suitable for click chemistry and can be used as molecular probes in vitro and in vivo. Studies indicate that (Z)-JQ1-TCO does not induce significant degradation of BRD4 in HeLa cells at 10 μM for 18 h.
  • Derivative of JQ1, an inhibitor of BET
  • Suitable for click chemistry
  • Can be used as molecular probes in vitro and in vivo
  • Does not induce significant degradation of BRD4 in HeLa cells at 10 μM for 18 h
  • Targets BRD4 and the Epigenetic Reader Domain