Learn More
Enzo Life Sciences Z-Leu-Leu-Tyr-ketoaldehyde (5 mg)
SDP
Supplier:  Enzo Life Sciences BMLZW86550005
Encompass_Preferred
A ketoaldehyde. An excellent inhibitor of the chymotrypsin-like activity of the proteasome and serine and cysteine proteases in general. Long Term Storage: -20°C.
Catalog No. 50-201-2379