Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Z-L-Phe Chloromethyl Ketone

Catalog No. 8607941G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-079-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 86-079-41G Supplier Sigma Aldrich Supplier No. 8607941G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Z-L-Phe Chloromethyl Ketone

Specifications

CAS 206996-57-8
Molecular Formula C5H12N4O3 -+ H2SO4 -+ H2O
Linear Formula H2NC(=NH)NHOCH2CH2CH(NH2)CO2H-+H2SO4-+H2O
Quantity 1 g
MDL Number MFCD00150048
Synonym N-Carbobenzyloxy-L-phenylalanyl chloromethyl ketone; NSC 251810; SL-01; ZPCK
Molecular Weight (g/mol) 292.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.