Learn More
Medchemexpress LLC (Z)-Lanoconazole | 101529-65-1 | 98.7% | C14H10ClN3S2 | 25 MG
SDP
Catalog No. 5000356495
Encompass_Preferred
(Z)-Lanoconazole is the Z configuration of Lanoconazole, a potent and orally active imidazole antifungal agent. It exhibits a broad spectrum of activity against fungi both in vitro and in vivo by interfering with ergosterol biosynthesis, specifically inhibiting sterol 14-alpha demethylase and blocking fungal membrane ergosterol biosynthesis. This agent is suitable for the investigation of dermatophytosis and onychomycosis.
  • Potent and orally active imidazole antifungal agent
  • Broad spectrum of activity against fungi in vitro and in vivo
  • Interferes with ergosterol biosynthesis
  • Inhibits sterol 14-alpha demethylase
  • Blocks fungal membrane ergosterol biosynthesis
  • Used for investigation of dermatophytosis and onychomycosis
  • Suppresses TPA-induced irritant dermatitis in vivo
  • Inhibits C. neoformans in vivo and reduces its growth in lungs and brains of MAIDS mice
Catalog No. 50-003-56495 Supplier Medchemexpress LLC Supplier No. 25MGHY14282A
(Z)-Lanoconazole is the Z configuration of Lanoconazole, a potent and orally active imidazole antifungal agent. It exhibits a broad spectrum of activity against fungi both in vitro and in vivo by interfering with ergosterol biosynthesis, specifically inhibiting sterol 14-alpha demethylase and blocking fungal membrane ergosterol biosynthesis. This agent is suitable for the investigation of dermatophytosis and onychomycosis.
  • Potent and orally active imidazole antifungal agent
  • Broad spectrum of activity against fungi in vitro and in vivo
  • Interferes with ergosterol biosynthesis
  • Inhibits sterol 14-alpha demethylase
  • Blocks fungal membrane ergosterol biosynthesis
  • Used for investigation of dermatophytosis and onychomycosis
  • Suppresses TPA-induced irritant dermatitis in vivo
  • Inhibits C. neoformans in vivo and reduces its growth in lungs and brains of MAIDS mice