Learn More
Medchemexpress LLC (Z)-lanoconazole | 101529-65-1 | MFCD00865590 | 99.3% | 319.8 g/mol | C14H10ClN3S2 | 5 MG
SDP
Catalog No. 5000221712
Encompass_Preferred
(Z)-Lanoconazole is an imidazole-class antifungal research compound (CAS 101529-65-1) supplied for laboratory use. It is provided as a high-purity solid and as pre-formulated DMSO solutions for in vitro antifungal assays and biochemical studies. Typical package sizes include low-milligram vials suitable for research workflows.
  • Imidazole-class antifungal compound for research applications.
  • High purity (~99.3%) appropriate for analytical and biological assays.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Supplied in small-scale vials for experimental workflows.
  • Product documentation includes COA and SDS for quality and safety.
Catalog No. 50-002-21712 Supplier Medchemexpress LLC Supplier No. HY14282A5MG
(Z)-Lanoconazole is an imidazole-class antifungal research compound (CAS 101529-65-1) supplied for laboratory use. It is provided as a high-purity solid and as pre-formulated DMSO solutions for in vitro antifungal assays and biochemical studies. Typical package sizes include low-milligram vials suitable for research workflows.
  • Imidazole-class antifungal compound for research applications.
  • High purity (~99.3%) appropriate for analytical and biological assays.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Supplied in small-scale vials for experimental workflows.
  • Product documentation includes COA and SDS for quality and safety.