Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-lanoconazole | 101529-65-1 | MFCD00865590 | 99.3% | C14H10ClN3S2 | 10MG
SDP

Supplier:  Medchemexpress LLC HY14282A10MG

Encompass_Preferred

(Z)-Lanoconazole is the Z-isomer of lanoconazole, an imidazole antifungal research compound that inhibits sterol 14-alpha demethylase and disrupts ergosterol biosynthesis. It is used in preclinical studies of dermatophytosis and onychomycosis and is supplied as a high-purity research chemical.

  • High purity (99.3%).
  • Molecular formula C14H10ClN3S2, molecular weight 319.83 g·mol⁻¹.
  • Available as powder and as a 10 mM solution in DMSO.
  • Soluble in DMSO at 100 mg/mL; documented in vivo formulation examples.
  • Recommended storage: powder -20°C (long term) or 4°C; in solvent -80°C or -20°C.

Catalog No. 50-002-61214


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.