Learn More
Enzo Life Sciences Z-LE(OMe)HD(OMe)-FMK (1mg)
SDP
Catalog No. 502008290
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-9. The carboxyl groups are OMe ester form to enhance cell permeability. If purified enzyme is used, esterase should be added for the hydrolysis of the ester groups. Alternative name: Caspase 9 inhibitor (fluoromethylketone). Formula: C32H43FN6O10. MW: 690.7. Purity: ≥95% (HPLC). Appearance: Off-white to tan solid. Formulation: TFA salt. Solubility: Soluble in DMSO (20mM). Long Term Storage: -20°C. Use/Stability: Stock solutions should be stored in aliquots at -20°C. Handling: Very hygroscopic.
Catalog No. 50-200-8290 Supplier Enzo Life Sciences Supplier No. ALX260076M001
Irreversible, cell permeable inhibitor of caspase-9. The carboxyl groups are OMe ester form to enhance cell permeability. If purified enzyme is used, esterase should be added for the hydrolysis of the ester groups. Alternative name: Caspase 9 inhibitor (fluoromethylketone). Formula: C32H43FN6O10. MW: 690.7. Purity: ≥95% (HPLC). Appearance: Off-white to tan solid. Formulation: TFA salt. Solubility: Soluble in DMSO (20mM). Long Term Storage: -20°C. Use/Stability: Stock solutions should be stored in aliquots at -20°C. Handling: Very hygroscopic.