Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-LEHD-FMK | 210345-04-3 | 98.09% | 690.72 | 25 MG
SDP

Catalog No. 5000426877
Encompass_Preferred

Z-LEHD-FMK is a selective and irreversible inhibitor of caspase-9. It protects against lethal reperfusion injury and attenuates apoptosis. It also shows a neuroprotective effect in a rat model of spinal cord trauma.

  • Selective and irreversible inhibitor of caspase-9.
  • Protects against lethal reperfusion injury.
  • Attenuates apoptosis.
  • Shows neuroprotective effect in a rat model of spinal cord trauma.
  • Purity of 98.09%.
  • Molecular weight is 690.72.
  • Appears as a white to yellow solid.

Catalog No. 50-004-26877 Supplier Medchemexpress LLC Supplier No. 25MGHYP1010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-LEHD-FMK is a selective and irreversible inhibitor of caspase-9. It protects against lethal reperfusion injury and attenuates apoptosis. It also shows a neuroprotective effect in a rat model of spinal cord trauma.

  • Selective and irreversible inhibitor of caspase-9.
  • Protects against lethal reperfusion injury.
  • Attenuates apoptosis.
  • Shows neuroprotective effect in a rat model of spinal cord trauma.
  • Purity of 98.09%.
  • Molecular weight is 690.72.
  • Appears as a white to yellow solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.