Learn More
Medchemexpress LLC Z-LEHD-FMK | 210345-04-3 | 98.1% | 690.72 | 10 MG
SDP
Catalog No. 5000422493
Encompass_Preferred
Z-LEHD-FMK is a selective and irreversible inhibitor of caspase-9. It protects against lethal reperfusion injury and attenuates apoptosis. This compound has also demonstrated neuroprotective effects in a rat model of spinal cord trauma.
  • Selective and irreversible inhibitor of caspase-9.
  • Protects against lethal reperfusion injury.
  • Attenuates apoptosis.
  • Exhibits neuroprotective effects in spinal cord trauma models.
Catalog No. 50-004-22493 Supplier Medchemexpress LLC Supplier No. HYP101010MG
Z-LEHD-FMK is a selective and irreversible inhibitor of caspase-9. It protects against lethal reperfusion injury and attenuates apoptosis. This compound has also demonstrated neuroprotective effects in a rat model of spinal cord trauma.
  • Selective and irreversible inhibitor of caspase-9.
  • Protects against lethal reperfusion injury.
  • Attenuates apoptosis.
  • Exhibits neuroprotective effects in spinal cord trauma models.