Learn More
Selleck Chemical LLC Z-LEHD-FMK TFA 2mg 524746-03-0 Caspase-9 Inhibitor
SDP
Catalog No. 501948139
Encompass_Preferred
Z-LEHD-FMK TFA (Caspase-9 Inhibitor) is a cell-permeable, competitive and irreversible inhibitor of enzyme caspase-9, which helps in cell survival. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8139 Supplier Selleck Chemical LLC Supplier No. S73132MG
Z-LEHD-FMK TFA (Caspase-9 Inhibitor) is a cell-permeable, competitive and irreversible inhibitor of enzyme caspase-9, which helps in cell survival. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.