Learn More
Selleck Chemical LLC Z-LEHD-FMK TFA
SDP
Catalog No. 5000004280
Encompass_Preferred
Z-LEHD-FMK TFA (Caspase-9 Inhibitor) is a cell-permeable competitive and irreversible inhibitor of enzyme caspase-9 which helps in cell survival
Catalog No. 50-000-04280 Supplier Selleck Chemical LLC Supplier No. S731310MG
Z-LEHD-FMK TFA (Caspase-9 Inhibitor) is a cell-permeable competitive and irreversible inhibitor of enzyme caspase-9 which helps in cell survival