Learn More
Medchemexpress LLC Z-leu-arg-7-amino-4-methylcoumarin | 156192-32-4 | MFCD00155595 | 99.2% | C30H38N6O6 | 10MG
SDP
Catalog No. 5000261369
Encompass_Preferred
Z-Leu-Arg-AMC is a benzyloxycarbonyl (Z)-protected leucyl-arginine peptide conjugated to 7-amino-4-methylcoumarin (AMC). It is a fluorogenic peptide substrate that releases the fluorescent AMC reporter upon proteolytic cleavage and is used to measure cysteine protease activity in biochemical assays.
Catalog No. 50-002-61369 Supplier Medchemexpress LLC Supplier No. HY14202110MG
Z-Leu-Arg-AMC is a benzyloxycarbonyl (Z)-protected leucyl-arginine peptide conjugated to 7-amino-4-methylcoumarin (AMC). It is a fluorogenic peptide substrate that releases the fluorescent AMC reporter upon proteolytic cleavage and is used to measure cysteine protease activity in biochemical assays.