Learn More
AdipoGen Z-Leu-Arg-Gly-Gly-AMC
SDP
Catalog No. 502562436
Encompass_Preferred
Chemical. CAS 167698-68-2 free base. Formula C34H44N8O8 . C2HF3O2. MW 692.8 . 114.0. Fluorogenic tetrapeptide substrate for ubiquitin C-terminal hydrolases UCHs, e.g. UCHL3 and isopeptidase T. LRGG is the preferred substrate sequence of the human deSUMOylating enzymes SENP6 and SENP7. Excitation 380nm. Emission 460nm.
Catalog No. 50-256-2436 Supplier AdipoGen Supplier No. AGCP30023M005
Chemical. CAS 167698-68-2 free base. Formula C34H44N8O8 . C2HF3O2. MW 692.8 . 114.0. Fluorogenic tetrapeptide substrate for ubiquitin C-terminal hydrolases UCHs, e.g. UCHL3 and isopeptidase T. LRGG is the preferred substrate sequence of the human deSUMOylating enzymes SENP6 and SENP7. Excitation 380nm. Emission 460nm.