Learn More
Enzo Life Sciences Z-Leu-Arg-Gly-Gly-AMC (5mg)
SDP
Catalog No. 502011368
Encompass_Preferred
Fluorogenic substrate for ubiquitin C-terminal hydrolases (UCHs), including UCH-L3 and Isopeptidase T. Sequence is based on the C-terminal residues of ubiquitin. Ex.: 380 nm Em.: 460 nm. Alternative name: Z-LRGG-AMC, Ubiquitin C terminal hydrolase substrate (fluorogenic), UCH substrate (fluorogenic). Purity: ≥95%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1368 Supplier Enzo Life Sciences Supplier No. BMLP8010005
Fluorogenic substrate for ubiquitin C-terminal hydrolases (UCHs), including UCH-L3 and Isopeptidase T. Sequence is based on the C-terminal residues of ubiquitin. Ex.: 380 nm Em.: 460 nm. Alternative name: Z-LRGG-AMC, Ubiquitin C terminal hydrolase substrate (fluorogenic), UCH substrate (fluorogenic). Purity: ≥95%. Solubility: Soluble in DMSO. Long Term Storage: -20°C.