Learn More
Enzo Life Sciences Z-Leu-Leu-CHO (25mg)
SDP
Supplier:  Enzo Life Sciences BMLPI1160025
Encompass_Preferred
Selective inhibitor of calpain, 100-fold selective for calpain over proteasome. Purity: ≥98% (TLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-1455