Learn More
AdipoGen Z-Leu-Leu-Glu-AMC
SDP
Catalog No. 502562438
Encompass_Preferred
Chemical. CAS 348086-66-8. Formula C35H44N4O9. MW 664.8. Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing PGPH caspase-like activity of the 20S proteasome. Excitation 380nm. Emission 460nm.
Catalog No. 50-256-2438 Supplier AdipoGen Supplier No. AGCP30022M005
Chemical. CAS 348086-66-8. Formula C35H44N4O9. MW 664.8. Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing PGPH caspase-like activity of the 20S proteasome. Excitation 380nm. Emission 460nm.