Learn More
Medchemexpress LLC Z-leu-leu-glu-AMC | 348086-66-8 | MFCD01074989 | 99.5% | 664.75 | C35H44N4O9 | 10MG
SDP
Catalog No. 5000237911
Encompass_Preferred
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to quantify the post-acidic (peptidylglutamyl) hydrolyzing activity of the 20S proteasome. Proteolytic cleavage releases the 7-amino-4-methylcoumarin (AMC) fluorophore, enabling sensitive fluorescence-based assays for biochemical research and enzyme activity screening.
  • Fluorogenic substrate that releases AMC upon proteolysis.
  • Suitable for measuring peptidylglutamyl-peptide hydrolyzing (PGPH/caspase-like) proteasome activity.
  • High reported purity for reliable assay results.
  • Available as lyophilized solid and as solution in DMSO for easy reconstitution.
  • Characterized by molecular weight 664.75 and formula C35H44N4O9.
Catalog No. 50-002-37911 Supplier Medchemexpress LLC Supplier No. HY12305310MG
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to quantify the post-acidic (peptidylglutamyl) hydrolyzing activity of the 20S proteasome. Proteolytic cleavage releases the 7-amino-4-methylcoumarin (AMC) fluorophore, enabling sensitive fluorescence-based assays for biochemical research and enzyme activity screening.
  • Fluorogenic substrate that releases AMC upon proteolysis.
  • Suitable for measuring peptidylglutamyl-peptide hydrolyzing (PGPH/caspase-like) proteasome activity.
  • High reported purity for reliable assay results.
  • Available as lyophilized solid and as solution in DMSO for easy reconstitution.
  • Characterized by molecular weight 664.75 and formula C35H44N4O9.