Learn More
Medchemexpress LLC Z-leu-leu-glu-AMC | 348086-66-8 | MFCD01074989 | 99.9% | 664.75 | C35H44N4O9 | 10MM 1ML
SDP
Catalog No. 5000237339
Encompass_Preferred
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure postacidic-like (peptidylglutamyl-peptide-hydrolyzing) activity of the 20S proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for proteasome activity assays, kinetic studies, and inhibitor screening. The product is available as a ready-to-use solution in DMSO and in multiple solid pack sizes for custom preparations.
  • Fluorogenic substrate that releases AMC upon cleavage.
  • Sensitive assay readout suitable for kinetic measurements.
  • Provided as ready-to-use 10 mM solution in DMSO or as solids for custom preparation.
  • High purity confirmed by LCMS.
  • Compatible with 20S and 26S proteasome activity assays.
Catalog No. 50-002-37339 Supplier Medchemexpress LLC Supplier No. HY12305310MM 1ML
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure postacidic-like (peptidylglutamyl-peptide-hydrolyzing) activity of the 20S proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for proteasome activity assays, kinetic studies, and inhibitor screening. The product is available as a ready-to-use solution in DMSO and in multiple solid pack sizes for custom preparations.
  • Fluorogenic substrate that releases AMC upon cleavage.
  • Sensitive assay readout suitable for kinetic measurements.
  • Provided as ready-to-use 10 mM solution in DMSO or as solids for custom preparation.
  • High purity confirmed by LCMS.
  • Compatible with 20S and 26S proteasome activity assays.