Learn More
Medchemexpress LLC Z-Leu-Leu-Glu-AMC | 348086-66-8 | MFCD01074989 | 99.5% | 664.75 | C35H44N4O9 | 25MG
SDP
Catalog No. 5000236327
Encompass_Preferred
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure postacidic-like (peptidylglutamyl) hydrolyzing activity of the 20S proteasome. Proteolytic cleavage releases the fluorescent 7-amino-4-methylcoumarin (AMC) reporter, enabling quantitative, sensitive measurement of proteasome activity in biochemical and enzymatic assays.
  • Fluorogenic substrate for proteasome activity assays.
  • Releases AMC reporter for sensitive fluorescence detection.
  • Suitable for biochemical and enzymatic assay development.
  • High purity for reproducible assay performance.
  • Available in small research-scale quantities.
Catalog No. 50-002-36327 Supplier Medchemexpress LLC Supplier No. HY12305325MG
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure postacidic-like (peptidylglutamyl) hydrolyzing activity of the 20S proteasome. Proteolytic cleavage releases the fluorescent 7-amino-4-methylcoumarin (AMC) reporter, enabling quantitative, sensitive measurement of proteasome activity in biochemical and enzymatic assays.
  • Fluorogenic substrate for proteasome activity assays.
  • Releases AMC reporter for sensitive fluorescence detection.
  • Suitable for biochemical and enzymatic assay development.
  • High purity for reproducible assay performance.
  • Available in small research-scale quantities.