Learn More
Medchemexpress LLC Z-leu-leu-glu-AMC | 348086-66-8 | MFCD01074989 | 99.5% | 664.75 g/mol | C35H44N4O9 | 1MG
SDP
Catalog No. 5000238680
Encompass_Preferred
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure post-acidic (caspase-like) hydrolytic activity of the 20S proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for kinetic assays, inhibitor screening, and proteasome activity profiling in biochemical research.
  • High purity (≈99.5%) for reliable assay performance.
  • Releases AMC for sensitive fluorometric detection.
  • Suitable for 20S/26S proteasome activity and inhibitor assays.
  • Available as solid and 10 mM DMSO solution formats.
  • Supplied in small research pack sizes for convenient use.
Catalog No. 50-002-38680 Supplier Medchemexpress LLC Supplier No. HY1230531MG
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure post-acidic (caspase-like) hydrolytic activity of the 20S proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for kinetic assays, inhibitor screening, and proteasome activity profiling in biochemical research.
  • High purity (≈99.5%) for reliable assay performance.
  • Releases AMC for sensitive fluorometric detection.
  • Suitable for 20S/26S proteasome activity and inhibitor assays.
  • Available as solid and 10 mM DMSO solution formats.
  • Supplied in small research pack sizes for convenient use.